Skip to Content
Merck
All Photos(1)

Key Documents

AOBH990FF8DD

AOBChem

4-Fluoro-3-(pyrrolidinomethyl)phenylboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
305.2
MDL number:

Iupac Name

1-(2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)pyrrolidine

Inchi Code

1S/C17H25BFNO2/c1-16(2)17(3,4)22-18(21-16)14-7-8-15(19)13(11-14)12-20-9-5-6-10-20/h7-8,11H,5-6,9-10,12H2,1-4H3

InChI key

PNOQVKHNDPYLQZ-UHFFFAOYSA-N

Purity

95

Country of Origin

CN

Product Link

https://aobchem.com/4-fluoro-3-pyrrolidinomethyl-phenylboronic-acid-pinacol-ester-44173.html

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library