Skip to Content
Merck
All Photos(1)

Key Documents

AMBH9884CCEE

Ambeed, Inc.

(4-Vinylphenyl)methanamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
133.19
MDL number:

Iupac Name

(4-vinylphenyl)methanamine

Inchi Code

1S/C9H11N/c1-2-8-3-5-9(7-10)6-4-8/h2-6H,1,7,10H2

InChI key

WAVDSLLYAQBITE-UHFFFAOYSA-N

storage temp.

Keep in dark place,Inert atmosphere,Store in freezer, under -20C

Purity

92%

Country of Origin

CN

Shipping Temperature

Cold-chain transportation

Physical Form

Colorless to yellow Liquid or semi-solid or Solid or lump

Product Link

https://www.ambeed.com/products/50325-49-0.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library