Iupac Name
3-bromo-4-methylpyridazine
Inchi Code
1S/C5H5BrN2/c1-4-2-3-7-8-5(4)6/h2-3H,1H3
InChI key
JBMFDMSMPJLATB-UHFFFAOYSA-N
storage temp.
Inert atmosphere,Store in freezer, under -20C
Purity
95%
Country of Origin
CN
Shipping Temperature
Cold-chain transportation
Physical Form
Solid or liquid
Product Link
https://www.ambeed.com/products/1416373-61-9.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.