Merck
All Photos(5)
Documents
BAF369740600
SP Bel-Art® Sterileware Powder Lance Samplers
white high-density polyethylene, capacity 125 mL (4 oz), L 23.625 in. (60 cm), sterile, pack of 20 ea (individually wrapped)
Synonym(s):
sampler
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
material
white high-density polyethylene
sterility
sterile
packaging
pack of 20 ea (individually wrapped)
manufacturer/tradename
Bel-Art F36974-0600
L
23.625 in. (60 cm)
capacity
125 mL (4 oz)
General description
A simple insert, rotate, and withdraw motion allows cross sectional sampling of non-flowing, bulk powders. Available in three lengths.
Features and Benefits
A simple insert, rotate, and withdraw motion allows cross sectional sampling of non-flowing, bulk powders. Available in three lengths.
  • Open blade works well with sticky, cohesive powders
  • Rounded edges help hold sample in place
  • Made from FDA and EU
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP