Skip to Content
Merck
All Photos(1)

Documents

900848

Sigma-Aldrich

4-Amino-2-trimethylsilanylethynyl-benzenesulfonyl fluoride

Synonym(s):

Amino sulfonyl fluoride TMS-alkyne, Probe building block

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14FNO2SSi
CAS Number:
Molecular Weight:
271.38
UNSPSC Code:
12352200
NACRES:
NA.22

form

liquid

availability

available only in USA

storage temp.

−20°C

SMILES string

NC1=CC=C(S(=O)(F)=O)C(C#C[Si](C)(C)C)=C1

Application

Used for chemical probe synthesis, this trifunctional building block contains an aryl sulfonyl fluoride group, alkyne tag, and amine synthetic handle. When appended to a ligand or pharmacophore through its amine linker, this building block allows for SuFEx-enabled, context-specific covalent modification of a biological target with potential for downstream applications via the alkyne tag. Use alone or in parallel with other multi-functional building blocks to discover the optimal probe for your chemical biology experiments.

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service