Skip to Content
Merck
All Photos(1)

Key Documents

724432

Sigma-Aldrich

Methyl-trioctylammonium bis(trifluoromethylsulfonyl)imide

Synonym(s):

NSC 747259, Methyltri-n-octylammonium bis(trifluoromethanesulfonyl)imide, Trioctylmethylammonium bis(trifluoromethylsulfonyl)imide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H54F6N2O4S2
CAS Number:
Molecular Weight:
648.85
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

liquid

refractive index

n20/D 1.439

density

1.095 g/mL at 25 °C

application(s)

battery manufacturing

SMILES string

FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC

InChI

1S/C25H54N.C2F6NO4S2/c1-5-8-11-14-17-20-23-26(4,24-21-18-15-12-9-6-2)25-22-19-16-13-10-7-3;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h5-25H2,1-4H3;/q+1;-1

InChI key

LAGQNGWYNLUQRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Capacitor technology, light-emitting devices, thin-film transistors, optical storage media

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vineet Kumar et al.
Bioorganic & medicinal chemistry letters, 19(16), 4643-4646 (2009-07-21)
The anti-cancer activity and cytotoxicity of phosphonium and ammonium-based ionic liquids (ILs) have been determined for the first time via NCI's in vitro 60 human tumor cell lines. The preliminary SAR showed that the chain length of alkyl substitution on
Lagrost, C.; Carrie, D.; Vaultier, M.; Hapiot, P.
The Journal of Physical Chemistry A, 107, 745-745 (2003)
Islam, Md. Mominul; Alam, Muhammad Tanzirul; Okajima, Takeyoshi; Ohsaka, Takeo
The Journal of Physical Chemistry C, 113, 3386-3386 (2009)
Koichi Iwata et al.
The journal of physical chemistry. B, 111(18), 4914-4919 (2007-04-13)
Time-resolved fluorescence spectra and fluorescence anisotropy decay of 2-aminoquinoline (2AQ) have been measured in eight room-temperature ionic liquids, including five imidazolium-based aromatic ionic liquids and three nonaromatic ionic liquids. The same experiments have also been carried out in several ordinary

Articles

Ionic liquids, also called room temperature ionic liquids, are organic salts that are liquid at, or close to, room temperature.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Lithium-ion batteries offer high energy density and cyclic performance for portable electronic devices.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service