Skip to Content
Merck
All Photos(1)

Key Documents

658596

Sigma-Aldrich

7-Chloroquinoline-4-boronic acid pinacol ester

90%

Synonym(s):

7-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17BClNO2
CAS Number:
Molecular Weight:
289.56
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

90%

form

solid

impurities

≤10 wt. % 7-Chloroquinoline

mp

81-87 °C

functional group

chloro

SMILES string

CC1(C)OB(OC1(C)C)c2ccnc3cc(Cl)ccc23

InChI

1S/C15H17BClNO2/c1-14(2)15(3,4)20-16(19-14)12-7-8-18-13-9-10(17)5-6-11(12)13/h5-9H,1-4H3

InChI key

GZDUYRAAWRRVEY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service