Skip to Content
Merck
All Photos(1)

Key Documents

635340

Sigma-Aldrich

PAMAM-tris(hydroxymethyl)amidomethane dendrimer, 1,4-diaminobutane core, generation 2 solution

10 wt. % in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)4NH2]:(G=2);dendri PAMAM[NHC(CH2OH)3]16
Molecular Weight:
4260.82
MDL number:
UNSPSC Code:
12162002

concentration

10 wt. % in methanol

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Contains 16 surface tris(hydroxymethyl)aminomethane groups.
The tris(hydroxymethyl)amido surface is an amplified neutral surface based on the trifunctional alcohol (tris hydroxyyl methylamine). This material offers a more densely alcohol functionalized particle. The neutral alcohol surface groups will be useful where more neutral pH conditions are necessary.

Legal Information

Mfg. by Dendritic NanoTechnologies Inc. Licensed for research use only.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service