Skip to Content
Merck
All Photos(1)

Documents

590118

Sigma-Aldrich

Ethyl 2-propylacrylate

99%, contains 150 ppm BHT as inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O2
CAS Number:
Molecular Weight:
142.20
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

contains

150 ppm BHT as inhibitor

refractive index

n20/D 1.4234 (lit.)

bp

141 °C (lit.)

density

0.880 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]\C([H])=C(/CCC)C(=O)OCC

InChI

1S/C8H14O2/c1-4-6-7(3)8(9)10-5-2/h3-6H2,1-2H3

InChI key

MXUMJFMINXROCH-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

115.0 °F - closed cup

Flash Point(C)

46.1 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service