Skip to Content
Merck
All Photos(1)

Documents

577189

Sigma-Aldrich

2-Naphthyl acrylate

90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10O2
CAS Number:
Molecular Weight:
198.22
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

90%

form

solid

mp

38-42 °C (lit.)

λmax

220 nm

SMILES string

C=CC(=O)Oc1ccc2ccccc2c1

InChI

1S/C13H10O2/c1-2-13(14)15-12-8-7-10-5-3-4-6-11(10)9-12/h2-9H,1H2

InChI key

SLVJUZOHXPZVLR-UHFFFAOYSA-N

Application

Fluorescent monomer

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service