Skip to Content
Merck
All Photos(1)

Key Documents

465623

Sigma-Aldrich

N-Methyl-N-phenylbenzotriazolemethanamine, mixture of Bt1 and Bt2 isomers

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N4
CAS Number:
Molecular Weight:
238.29
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

reaction suitability

reaction type: C-C Bond Formation

mp

58-70 °C (lit.)

SMILES string

CN(Cn1nnc2ccccc12)c3ccccc3

InChI

1S/C14H14N4/c1-17(12-7-3-2-4-8-12)11-18-14-10-6-5-9-13(14)15-16-18/h2-10H,11H2,1H3

InChI key

VIBLZNXJYGXWHR-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service