Skip to Content
Merck
All Photos(1)

Key Documents

AC7300013742

Advanced Chemical Intermediates Ltd.

2,3-Dihydro-benzofuran-2-carbonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
182.61
MDL number:

Iupac Name

2,3-dihydro-1-benzofuran-2-carbonyl chloride

Inchi Code

1S/C9H7ClO2/c10-9(11)8-5-6-3-1-2-4-7(6)12-8/h1-4,8H,5H2

InChI key

QZVPHVMVWZKULP-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Room temperature

Physical Form

Colourless oil

Product Link

https://www.acints.com/products/ACI-01144

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

MSDS

https://www.acints.com/products/ACI-01144


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library