Merck
  • Design, synthesis and biological evaluation of chalconyl blended triazole allied organosilatranes as giardicidal and trichomonacidal agents.
Design, synthesis and biological evaluation of chalconyl blended triazole allied organosilatranes as giardicidal and trichomonacidal agents.
European journal of medicinal chemistry (2015-12-24)
Gurjaspreet Singh, Aanchal Arora, Satinderpal Singh Mangat, Sunita Rani, Hargobinder Kaur, Kapil Goyal, Rakesh Sehgal, Indresh Kumar Maurya, Rupinder Tewari, Duane Choquesillo-Lazarte, Subash Sahoo, Navneet Kaur
ABSTRACT
A series of chalconyl blended triazole allied silatranes (7a-g/8a-g/9a-g) were synthesized in good yields using a simple, economical and biocompatible synthetic route. The blend of three different pharmacologically active moieties into a single scaffold resulted into synergistic effect in their bio-activity. Various substitutions were tried to study the structure activity relationship (SAR) of the synthesized compounds on the basis of biological results. All the newly synthesized compounds were well characterized by IR, (1)H and (13)C NMR, low resolution mass spectroscopy and elemental analysis. The structures of 7a and 7c were authenticated by single crystal X-ray crystallography. These compounds were screened by using Molinspiration software for their physicochemical properties and all the compounds showed good oral bioavailability. The antiparasitic activity of the newly synthesized compounds was evaluated against unicellular parasites (Giardia lamblia and Trichomonas vaginalis) in comparison to standard drug (metronidazole) by 3-(4,5-dimethylthiazol-yl)-diphenyl tetrazolium bromide (MTT) assay. All the compounds displayed significant activity against G. lamblia and T. vaginalis with IC50 values ranging from 19.58-131.2 μM to 18.24-101.26 μM respectively. The entire library of compounds was found to be more active than metronidazole except 9a, 9f and 9g. Notably, 9e and 7e were found to be most significant against G. lamblia and T. vaginalis respectively.
Sigma-Aldrich
Bromotris(triphenylphosphine)copper(I), 98%