MilliporeSigma
  • Implementation of mixture design for formulation of albumin containing enteric-coated spray-dried microparticles.
Implementation of mixture design for formulation of albumin containing enteric-coated spray-dried microparticles.
Drug development and industrial pharmacy (2012-05-18)
Prathap Nagaraja Shastri, Ruhi V Ubale, Martin J D'Souza
ABSTRACT
Oral delivery of proteins has been a challenging as well as rapidly developing field. To implement mixture design of experiment to develop enteric-coated microparticles containing bovine serum albumin. Microparticles were prepared using Buchi Spray Dryer 191. Simplex lattice mixture design computed using JMP software was implemented to compare the gastric protection rendered by Eudragit FS30D, Eudragit L100-55, and Eudragit S100 in microparticulate form. Further, an extreme vertices mixture design was used to incorporate hydroxypropyl methylcellulose (HPMC) Chitosan in the formulation to delay the release. Microparticle recovery yield and protein content in microparticles were evaluated. The design was statistically significant with Eudragit S100 resulting in protein release of < 5% in acidic buffer. The selected optimal formulation had 70% of Eudragit S, 25% HPMC, and 5% Chitosan. The release profiles of protein from Eudragit S alone and along with HPMC were compared. About 25% decrease in the amount of protein release was observed 6 h post exposure of microparticle to buffer of pH 6.8. The microparticle recovery yield reduced from 77.99% to 71.56% which is due to addition of HPMC into the formulation matrix. Although all three Eudragit polymers can be used for enteric coating, in the microparticulate form Eudragit S resulted in higher gastric protection. Also use of HPMC along with Eudragit S resulted in further sustained release.
Sigma-Aldrich
Poly(methyl methacrylate-co-methacrylic acid), average Mw ~34,000 by GPC, average Mn ~15,000 by GPC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
500 g
In Stock
Details...
$193.00
$183.35
1 kg
In Stock
Details...
$331.00
$291.28
To order products, please contact your local dealer.