Merck
  • High throughput genome-specific and gene-specific molecular markers for erucic acid genes in Brassica napus (L.) for marker-assisted selection in plant breeding.
High throughput genome-specific and gene-specific molecular markers for erucic acid genes in Brassica napus (L.) for marker-assisted selection in plant breeding.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik (2008-07-18)
Mukhlesur Rahman, Zudong Sun, Peter B E McVetty, Genyi Li
ABSTRACT
A single base change in the Bn-FAE1.1 gene in the A genome and a two-base deletion in the Bn-FAE1.2 gene in the C genome produce the nearly zero content of erucic acid observed in canola. A BAC clone anchoring Bn-FAE1.1 from a B. rapa BAC library and a BAC clone anchoring Bn-FAE1.2 from a B. oleracea BAC library were used in this research. After sequencing the gene flanking regions, it was found that the dissimilarity of the flanking sequences of these two FAE1 homologs facilitated the design of genome-specific primers that could amplify the corresponding genome in allotetraploid B. napus. The two-base deletion in the C genome gene was detected as a sequence-characterized amplified region (SCAR) marker. To increase the throughput, one genome-specific primer was labeled with four fluorescence dyes and combined with 20 different primers to produce PCR products with different fragment sizes. Eventually, a super pool of 80 samples was detected simultaneously. This dramatically reduces the cost of marker detection. The single base change in the Bn-FAE1.1 gene was detected as single nucleotide polymorphic (SNP) marker with an ABI SNaPshot kit. A multiplexing primer set was designed by adding a polyT to the 5' primer end to increase SNP detection throughput through sample pooling. Furthermore, the Bn-FAE1.1 and Bn-FAE1.2 were integrated into the N8 and N13 linkage groups of our previously reported high-density sequence-related amplified polymorphism (SRAP) map, respectively. There were 124 SRAP markers in a N8 bin in which the Bn-FAE1.1 gene-specific SCAR marker was located and 46 SRAP markers in a N13 bin into which the Bn-FAE1.2 SNP marker was integrated. These three kinds of high throughput molecular markers have been successfully implemented in our canola/rapeseed breeding programs.
Sigma-Aldrich
Erucic acid, technical, ~90% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Erucic acid, ≥99% (capillary GC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Erucic acid, analytical standard