Merck
  • A naphthalene-based fluorescent probe with a large Stokes shift for mitochondrial pH imaging.
A naphthalene-based fluorescent probe with a large Stokes shift for mitochondrial pH imaging.
The Analyst (2018-09-22)
Bo Lin, Li Fan, Jinyin Ge, Wenjia Zhang, Caihong Zhang, Chuan Dong, Shaomin Shuang
ABSTRACT
A mitochondrial-targeted pH fluorescent probe 4-(2-(6-hydroxynaphthalen-2-yl)vinyl)-1-methylpyridin-1-ium was facilely synthesized via the carbon-carbon double bond bridging of 6-hydroxy-2-naphthaldehyde and 1,4-dimethylpyridinium iodide salt. The probe exhibited remarkable pH-dependent behavior in the linear range of 7.60-10.00, with a pKa value of 8.85 ± 0.04 near mitochondrial pH. A significantly large Stokes shift of 196 nm was obtained, which reduces the interference of excitation light. Application of the probe in live HepG2 cells indicated that the probe had excellent mitochondrial targeting ability and was successfully used to visualize mitochondrial pH fluctuations in live cells.
Sigma-Aldrich
6-hydroxy-2-naphthaldehyde, AldrichCPR
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1,3-Propylene sulfite, 99%