Skip to Content
Merck
All Photos(1)

Key Documents

CPAE

86111401, bovine cardiovascular , Endothelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

CPAE, 86111401

biological source

bovine cardiovascular

packaging

tube of 5 μg 86111401-DNA-5UG
pkg of vial of cells 86111401-1VL

growth mode

Adherent

karyotype

2n = 60, diploid

morphology

Endothelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Bovine pulmonary artery endothelium

Cell Line Description

Established from the main stem pulmonary artery of a young, female cow (Bos taurus). The cells have angiotensin-converting enzyme activity. BVDV positive.

Application

Enzymatic studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service