N1230010
Norfloxacin impurity A
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
7-Chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
About This Item
grade
pharmaceutical primary standard
API family
norfloxacin
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
Fc1cc2c(cc1Cl)N(C=C(C2=O)C(=O)[O-])CC
InChI
1S/C12H9ClFNO3/c1-2-15-5-7(12(17)18)11(16)6-3-9(14)8(13)4-10(6)15/h3-5H,2H2,1H3,(H,17,18)/p-1
InChI key
WNNSMMJBBOPPOT-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service