Skip to Content
Merck
All Photos(1)

Documents

BP090

Codeine phosphate

British Pharmacopoeia (BP) Assay Standard

Synonym(s):

Codeine phosphate hemihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21N2O3 · H3PO4 · 0.5H2O
CAS Number:
Molecular Weight:
420.37
UNSPSC Code:
41116107
NACRES:
NA.24

manufacturer/tradename

BP

drug control

estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Codeine phosphate BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Paracetamol, Codeine Phosphate and Caffeine Capsules
  • Co-dydramol Tablets
  • Co-codamol Capsules
  • Co-codamol Tablets
  • Co-codamol Effervescent Tablets
  • Paracetamol, Codeine Phosphate and Caffeine Tablets
  • Morphine Tablets
  • Morphine Prolonged-release Tablets
  • Dihydrocodeine Prolonged-release Tablets
  • Morphine Prolonged-release Capsules
  • Morphine Granules for Oral Suspension
  • Dihydrocodeine Oral Solution
  • Co-codaprin Tablets
  • Dihydrocodeine Injection
  • Morphine Oral Solution
  • Dihydrocodeine Tablets
  • Morphine Suppositories
  • Morphine Capsules
  • Morphine Sulfate Injection
  • Codeine Phosphate Oral Solution
  • Packaging

    Unit quantity: 200 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Pictograms

    Skull and crossbones

    Signal Word

    Danger

    Hazard Statements

    Hazard Classifications

    Acute Tox. 3 Oral

    Storage Class Code

    6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service