Key Documents
Z420220
Sigma-Aldrich
Spinbar® disposable magnetic stirring bar
size 1 in. × 5/16 in.
Synonym(s):
Stir Bar, Stirbar, Stirring Bar
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
Pricing and availability is not currently available.
material
PTFE-coated
polygon PTFE-coated
packaging
pkg of 100 ea
manufacturer/tradename
Bel-Art F37112-0001
size
1 in. × 5/16 in.
General description
For single-use application where efficiency and cross-contamination are a concern. Alnico V magnet.
Legal Information
Spinbar is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)