Key Documents
SYNF459050
Synthware Squibb-style separatory funnel with standard taper joints, PTFE stopcock and PTFE stopper
50 mL, joint: ST/NS 19/22, PTFE stopcock and PTFE stopper
Synonym(s):
Synthware F459050, glass separatory funnel
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F459050
capacity
50 mL
50 mL
General description
With 2 mm PTFE stopcock. Supplied with a glass stopper or PTFE stopper.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)