Skip to Content
Merck
All Photos(1)

Documents

F9000

Sigma-Aldrich

2-Fluoro-9-fluorenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H7FO
CAS Number:
Molecular Weight:
198.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

185 °C/10 mmHg (lit.)

mp

115-117 °C (lit.)

SMILES string

Fc1ccc2-c3ccccc3C(=O)c2c1

InChI

1S/C13H7FO/c14-8-5-6-10-9-3-1-2-4-11(9)13(15)12(10)7-8/h1-7H

InChI key

CNCFLCLXJBPMIR-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesco Ciriello et al.
Environmental science and pollution research international, 22(16), 12469-12478 (2015-04-24)
Airborne particulate matter (PM) contains several quinones, which are able to generate reactive oxygen species impacting on cell viability. A method able to detect and quantify PM oxidative potential, based on the cytochrome c (cyt-c) reduction by means of superoxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service