BR362138
BRAND® BLAUBRAND® ASTM 100 mL centrifuge tube
pear-shaped, cylindrical, former standard ASTM D96
Synonym(s):
ASTM, BLAUBRAND®, Centrifuge Tubes, Tubes
All Photos(1)
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
Recommended Products
material
borosilicate glass
cylindrical
pear-shaped
sterility
non-sterile
feature
RCF 700 × g
packaging
pack of 2
manufacturer/tradename
BRAND 362138
L
160 mm
Related Categories
General description
Withstands RCF up to 700. Design, accuracy, etc. to meet ASTM ("American Society for Testing and Materials") specifications.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)