Skip to Content
Merck
All Photos(1)

Key Documents

40955

Sigma-Aldrich

Methoxy(dimethyl)octadecylsilane

technical, ≥90% (GC)

Synonym(s):

Dimethylmethoxyoctadecylsilane, Dimethyloctadecylmethoxysilane, Octadecyldimethylmethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H46OSi
CAS Number:
Molecular Weight:
342.67
Beilstein:
7346074
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

technical

Assay

≥90% (GC)

form

solid

impurities

5-10% other C18-isomers

refractive index

n20/D 1.444 (lit.)
n20/D 1.444

density

0.83 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC[Si](C)(C)OC

InChI

1S/C21H46OSi/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-23(3,4)22-2/h5-21H2,1-4H3

InChI key

RUFRLNPHRPYBLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methoxy(dimethyl)octadecylsilane(MDOS) is an organosilane with one methoxy group. It is a silane coupling agent which forms a self-assembled monolayer(SAM) on a variety of particles.

Application

MDOS can be used as an adhesion promoter between wool fiber and the polyethylene matrix that can be used to conserve waste of wool and poultry feathers. Wood fibre silanized with MDOS can be used as reinforcement to prepare polyethylene based composites with improved thermo-oxidative stability.
Silica spheres for HPLC colums were reported to be functionalized with methoxy(dimethyl) octadecylsilane. 1 Methoxy(dimethyl) octadecylsilane may be grafted on the surface of ZnO nanoparticles. 2

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wool fibres functionalised with a silane-based coupling agent for reinforced polypropylene composites.
Conzatti L, et al.
Composites Part A: Applied Science and Manufacturing, 61, 51-59 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service