- Titin force enhancement following active stretch of skinned skeletal muscle fibres.
Titin force enhancement following active stretch of skinned skeletal muscle fibres.
The Journal of experimental biology (2017-06-24)
Krysta Powers, Venus Joumaa, Azim Jinha, Eng Kuan Moo, Ian Curtis Smith, Kiisa Nishikawa, Walter Herzog
PMID28637823
ABSTRACT
In actively stretched skeletal muscle sarcomeres, titin-based force is enhanced, increasing the stiffness of active sarcomeres. Titin force enhancement in sarcomeres is vastly reduced in