Skip to Content
Merck
All Photos(1)

Key Documents

SAAR5681100

Safranin O

Synonym(s):

Basic Red 2, Cotton Red, Gossypimine, Safranin T, Safranin Y or A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19ClN4
CAS Number:
Molecular Weight:
350.84
Colour Index Number:
50240
Beilstein:
3924099
MDL number:
UNSPSC Code:
41116100

packaging

poly bottle of 20 × 25 g (SAAR5681100CB)

storage temp.

2-30°C

SMILES string

[Cl-].Cc1cc2nc3cc(C)c(N)cc3[n+](-c4ccccc4)c2cc1N

InChI

1S/C20H18N4.ClH/c1-12-8-17-19(10-15(12)21)24(14-6-4-3-5-7-14)20-11-16(22)13(2)9-18(20)23-17;/h3-11H,1-2H3,(H3,21,22);1H

InChI key

OARRHUQTFTUEOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For laboratory use only

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service