Skip to Content
Merck
All Photos(1)

Documents

2-Methylisoborneol solution

analytical standard

Synonym(s):

(1R-exo)-1,2,7,7-Tetramethylbicyclo[2.2.1]heptan-2-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20O
CAS Number:
Molecular Weight:
168.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1 × 1 mL (47523-U)
ampule of 5 × 2 mL (4M7523-U)

concentration

100 μg/mL in methanol

application(s)

environmental

storage temp.

room temp

SMILES string

CC1(C)[C@@H]2CC[C@@]1(C)[C@](C)(O)C2

InChI

1S/C11H20O/c1-9(2)8-5-6-10(9,3)11(4,12)7-8/h8,12H,5-7H2,1-4H3/t8-,10-,11-/m1/s1

InChI key

LFYXNXGVLGKVCJ-FBIMIBRVSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service