Skip to Content
Merck
All Photos(1)

Key Documents

Z730432

Sigma-Aldrich

Biomaterials Science: An Introduction to Materials in Medicine, 2nd ed.

over 80 contributors from academia, government and industry

Sign Into View Organizational & Contract Pricing


About This Item

ISBN-13:
978-0-12-582463-7
UNSPSC Code:
55101509

publication info

B. Ratner, et al., ed., Elsevier, 2004, 864 pp., hard cover

manufacturer/tradename

Elsevier

General description

The second edition provides the most up-to-date comprehensive review of all aspects of biomaterials science. It integrates a historical perspective of materials engineering principles with biological interactions of biomaterials. Also provided within are regulatory and ethical issues in addition to future directions of the field, and a state-of-the-art update of medical and biotechnological applications.

All aspects of biomaterials science are thoroughly addressed, from tissue engineering to cochlear prostheses and drug delivery systems. Over 80 contributors from academia, government and industry detail the principles of cell biology, immunology, and pathology. Focus within pertains to the clinical uses of biomaterials as components in implants, devices, and artificial organs. This reference also touches upon their uses in biotechnology as well as the characterization of the physical, chemical, biochemical and surface properties of these materials.

Table of Contents

INTRODUCTION
BIOMATERIALS HISTORY

PART I: Materials Science & Engineering
Properties of Materials

Introduction
Bulk Properties of Materials
Finite Element Analysis
Surface Properties of Materials
Role of Water in Biomaterials

Classes of Materials Used in Medicine
Introduction
Metals
Polymers
A Brief History of Silicone Biomaterials
Hydrogels
Bioresorbable and Biodegradable Materials
Ceramics, Classes and Class-Ceramics
Natural Materials
Composites
Thin Films, Grafts, Coatings
Medical Fibers and Biotextiles
Biological Functional Materials
Smart Materials
Pyrolytic Carbon for Long-Term Medical Implants
Textured and Porous Materials
Non-fouling Surfaces

PART II: Biology, Biochemistry and Medicine
Background Concepts

Introduction
The Role of Absorbed Proteins in Tissue Response to Biomaterials
Calls and Cell Injury
Tissues: The Extracellular Matrix and Call-Biomaterials Interactions
Mechanical Forces on Cells

Host Reactions to Biomaterials and Their Evaluation
Testing Biomaterials

Introduction
Inflammation, Wound Healing and the Foreign Body Response
The Complement Systems
Systemic Toxicity and Hypersensitivity
Blood Coagulation and Blood-Materials Interactions
Tumorigenesis and Biomaterials
Infection

Testing Biomaterials
Introduction
In Vitro Assessment of Tissue Compatibility
In Vivo Assessment of Tissue Compatibility
Testing of Blood-Materials Interactions
Large Animal Models in Cardiac and Vascular Biomaterials Research and Testing
Microscopy for Biomaterials Science

Degradation of Materials in the Biological Environment
Introduction
Degradation of Materials in the Biological Environment
Degradative Effects of the Biological Environment on Metals and Ceramics
Pathologic Calcification of Biomaterials

Applications of Materials in Medicine, Dentistry and Biology
Introduction
Cardiovascular Medical Devices
Nonthrombogenic Treatments and Strategies
Dental Implantation
Adhesive and Sealants
Ophthalmologic Applications
Intraocular Lens Implants: A Scientific Perspective
Clinical Aspects of Orthopedic Biomaterial Performance
Drug Delivery Systems
Sutures
Burn Dressings and Skin Substitutes
Bioelectrodes
Biomedical Sensors and Biosensors
Diagnostics and Biomaterials

Artificial Organs and Tissues
Introduction
Implantable Cardiac Assist Devices
Extracorporeal Artificial Organs
Cochlear Implants
Artificial Red Blood Cell Substitutes

Tissue Engineering
Introduction
Overview of Tissue Engineering
Immunoissolation
Synthetic Bioresorbable Polymer Scaffolds

PART III: Practical Aspects of Biomaterials
Implants & Devices - Special Concerns

Introduction
Sterilization of Implants and Devices
Implants and Device Failure
Correlations of Materials Surface Properties with Biological Responses
Implant Retrieval and Evaluation

New Products and Standards
Introduction
Voluntary Consensus Standards
Development and Regulation of Medical Products Using Biomaterials
Ethical Issues in the Development of New Biomaterials
Legal Aspects of Biomaterials
Perspectives and Possibilities in Biomaterials Science
APPENDICES

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service