Skip to Content
Merck
All Photos(1)

Documents

VIV108011

SynVivo SynBBB 3D blood brain barrier model chip, IMN2 linear configuration

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
60104506
NACRES:
NB.35

packaging

case of 1 ea

manufacturer/tradename

SynVivo 108011

storage temp.

room temp

General description

Linear co-culture chip-IMN2-Linear Channel Barrier type: Slits, Slit width: 3 μm, Distance between slits: 50 μm, Travel/barrier width: 50 μm, Outer Channel width: 200 μm, Central Channel width: 500 μm, Device depth: 100 μm. This device enables a novel microfluidics based tubular Airway and Air Liquid Interface model mimicking the lung architecture.

Linkage

LInk to Publications
application text

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service