Skip to Content
Merck
All Photos(1)

Documents

SML1344

Sigma-Aldrich

AK-1

≥98% (HPLC)

Synonym(s):

3-[(Hexahydro-1H-azepin-1-yl)sulfonyl]-N-(3-nitrophenyl)-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21N3O5S
CAS Number:
Molecular Weight:
403.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

O=S(N1CCCCCC1)(C2=CC(C(NC3=CC=CC([N+]([O-])=O)=C3)=O)=CC=C2)=O

InChI

1S/C19H21N3O5S/c23-19(20-16-8-6-9-17(14-16)22(24)25)15-7-5-10-18(13-15)28(26,27)21-11-3-1-2-4-12-21/h5-10,13-14H,1-4,11-12H2,(H,20,23)

InChI key

HAYBKCHPEBZNGW-UHFFFAOYSA-N

Biochem/physiol Actions

AK-1 is a benzylsulfonamide. It acts as avia inhibiting of the nuclear factor-KB (NF-KB) /CSN2 (casein β) pathway.
AK-1 is a cell permeable, potent and specific SIRT2 inhibitor that induced proteasomal degradation of the Snail transcription factor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AK-1, a specific SIRT2 inhibitor, induces cell cycle arrest by downregulating Snail in HCT116 human colon carcinoma cells.
Cheon M G, et al.
Cancer Letters, 356(2), 637-645 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service