Skip to Content
Merck
All Photos(1)

Documents

SAB2702173

Sigma-Aldrich

Monoclonal Anti-FANCD2 antibody produced in mouse

Synonym(s):

FACD, FAD, FAD2, FANCD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

103
monoclonal

form

liquid

species reactivity

human

concentration

1 mg/mL

technique(s)

ELISA: suitable
western blot: 1:500-1:3000

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FANCD2(2177)

General description

Fanconi anemia complementation group D2 (FANCD2) is encoded by the gene mapped to human chromosome 3p25.3.

Immunogen

Human FANCD2 fusion protein

Application

For WB: Use at a dilution of 1:500-1:3000. For ELISA: Use at an assay dependent dilution. Optimal dilutions/concentrations should be determined by the researcher.

Biochem/physiol Actions

Fanconi Anemia Complementation Group D2 (FANCD2) protein is one of at least six factors shown to be involved in the autosomal-recessive cancer-prone disorder, Fanconi Anemia (FA). FA group D has been shown to be comprised of two separate proteins, FANCD1 and FANCD2. Mutations in BRCA2 can cause FANCD1. FANCD2 has been shown to colocalize with BRCA1 in ionizing radiation-induced foci. FANCD2 is involved in genomic resistance to DNA cross-linking reagents, and the arrest of DNA synthesis following exposure to ionizing radiation. FANCD2 has been shown to directly interact with NBS1
Fanconi anemia complementation group D2 (FANCD2) is implicated in countering endogenous levels of replication stress, which is required for maintenance of genomic stability. FANCD2 plays a crucial role in fanconi anemia (FA) pathway. It is also involved in the regulation of human papillomavirus (HPV) replication. Overexpression of the gene has been observed in hepatocellular carcinoma (HCC). Thus, it can be considered as a potential biomarker for poor prognosis and therapeutic target for HCC.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

FANCD2 Binds Human Papillomavirus Genomes and Associates with a Distinct Set of DNA Repair Proteins to Regulate Viral Replication
Spriggs CC and Laimins LA
mBio, 8, e02340-e02316 (2017)
Constitutive role of the Fanconi anemia D2 gene in the replication stress response.
Tian Y, et al.
The Journal of Biological Chemistry, 292, 20184-20195 (2017)
Clinical Significance of FANCD2 Gene Expression and its Association with Tumor Progression in Hepatocellular Carcinoma.
Komatsu H, et al.
Anticancer Research, 37, 1083-1090 (2017)
Localization of the Fanconi Anemia Complementation Group D Gene to a 200-kb Region on Chromosome 3p25.3
Hejna JA, et al.
American Journal of Human Genetics, 66, 1540-1551 (2000)
Joshi Niraj et al.
Nucleic acids research, 45(14), 8341-8357 (2017-07-02)
Fanconi anemia (FA) is a recessive genetic disorder characterized by congenital abnormalities, progressive bone-marrow failure, and cancer susceptibility. The FA pathway consists of at least 21 FANC genes (FANCA-FANCV), and the encoded protein products interact in a common cellular pathway

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service