S8629
Sulfatase from Patella vulgata (keyhole limpet)
Type V, essentially salt-free, lyophilized powder, ≥5 units/mg solid
Synonym(s):
Aryl-sulfatase, Aryl-sulfate sulfohydrolase, Phenolsulfatase
About This Item
Recommended Products
type
Type V
form
essentially salt-free, lyophilized powder
specific activity
≥5 units/mg solid
foreign activity
β-Glucuronidase ≤2 unit/mg solid
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Application
Unit Definition
substrate
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service