Skip to Content
Merck
All Photos(1)

Documents

S2941

Sigma-Aldrich

SKF-75670 hydrobromide

>98% (HPLC), solid

Synonym(s):

7,8-dihydroxy-3-methyl-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO2 · HBr
CAS Number:
Molecular Weight:
350.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

>98% (HPLC)

form

solid

color

white

solubility

DMSO: soluble 12 mg/mL
H2O: insoluble

SMILES string

Br.CN1CCc2cc(O)c(O)cc2C(C1)c3ccccc3

InChI

1S/C17H19NO2.BrH/c1-18-8-7-13-9-16(19)17(20)10-14(13)15(11-18)12-5-3-2-4-6-12;/h2-6,9-10,15,19-20H,7-8,11H2,1H3;1H

InChI key

KWTPHNVUAVFKGB-UHFFFAOYSA-N

Gene Information

human ... DRD1(1812)

Application

SKF-75670 may be used in dopamine and cocaine-mediated cell signaling studies.

Biochem/physiol Actions

SKF-75670 is a selective dopamine D1 receptor agonist but with lower efficacy. It also acts as cocaine antagonist and alters the behavioral effects of cocaine in squirrel monkeys.
Atypical D1 dopamine receptor agonist. Displays antagonist activity in vitro and agonist activity in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R D Spealman et al.
Psychopharmacology, 133(3), 283-292 (1997-11-15)
Dopamine D1 agonists differing in efficacy with respect to stimulation of adenylate cyclase activity and other in vitro and in vivo criteria were evaluated for their capacity to modulate the behavioral effects of cocaine in squirrel monkeys. Monkeys were trained
P H Andersen et al.
European journal of pharmacology, 188(6), 335-347 (1990-06-12)
Dopamine receptor selectivity was investigated for a number of dopamine receptor agonists. In vitro, the benzazepine derivatives, e.g., SKF 38393 and SKF 75670 as well as the isoquinoline derivatives, SKF 89626 and SKF 89615, were D1 receptor-selective. All other compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service