Skip to Content
Merck
All Photos(2)

Key Documents

MAK376

Sigma-Aldrich

Covidyte IF670 Substrate

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

usage

sufficient for 100 fluorometric tests

application(s)

pharmaceutical

detection method

fluorometric

relevant disease(s)

immunological diseases; pulmonary disorders

storage temp.

−20°C

General description

The coronavirus main protease, which plays a pivotal role in viral gene expression and replication through the proteolytic processing of replicase polyproteins, is an attractive target for anti-CoV drug design. The inhibition of viral proteases necessary for proteolytic processing of polyproteins has been a successful strategy in the treatment of human immunodeficiency virus (HIV) and hepatitis C respectively, proving the potential of protease inhibitors for the treatment of viral infections. Similarly, the main protease of SARS-CoV-2 is thought to be essential for viral replication and is therefore regarded as a promising target for antiviral therapy of Covid-19.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for developing methods to screen potential inhibitors of coronavirus proteases

Principle

Covidyte IF670 is a peptide substrate containing a 12-amino acid sequence (VNSTLQSGLRKM) that can be cleaved by coronavirus proteases. The dark-FRET peptide contains Tide Quencher 5 (TQ5) as a quencher and iFluor 670 as a fluorescent donor on the N- and C- terminals respectively where the fluorescence of iFluor 670 is effectively quenched by TQ5 when the peptide is intact. When the peptide is hydrolyzed by coronavirus proteases, the iFluor 670 fragment generates significantly enhanced fluorescence since its fluorescence is no longer quenched by TQ5. The activity of coronavirus proteases can be effectively monitored by the fluorescence intensity of iFluor 670.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Covidyte is a trademark of AAT Bioquest, Inc.
Tide Quencher is a trademark of AAT Bioquest, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service