Skip to Content
Merck
All Photos(1)

Key Documents

M4164

Sigma-Aldrich

MAN-5-Asn Glycan

~90%

Synonym(s):

(Man)5(GlcNAc)2As, Mannopentaose-di-(N-acetyl-D-glucosamine)-asparagine, Oligomannose-5-Asn glycan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H84N4O38
CAS Number:
Molecular Weight:
1349.21
MDL number:
UNSPSC Code:
12352201

Pricing and availability is not currently available.

Assay

~90%

storage temp.

−20°C

SMILES string

CC(=O)NC1C(O)C(OC2OC(CO)C(OC3OC(COC4OC(COC5OC(CO)C(O)C(O)C5O)C(O)C(OC6OC(CO)C(O)C(O)C6O)C4O)C(O)C(OC7OC(CO)C(O)C(O)C7O)C3O)C(O)C2NC(C)=O)C(CO)OC1NC(=O)CC(N)C(O)=O

Other Notes

N-Linked glyco-amino acid; fragment of ovalbumin glycoprotein.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service