EP022628124
Eppendorf® Centrifuge 5702/5702R/5702RH
5702RH, refrigerated/heated, with rotor A-4-38 and 4 x 85 mL round bucket, rotary knobs, AC/DC input 120 V, 60 Hz, US 3-pin plug (US)
About This Item
feature
CSA approved
to hold 4 100 mL round buckets
to hold 4 50 ml conical tubes
to hold 8 15 mL conical tubes
refrigerated/heated
rotary knobs
manufacturer/tradename
Eppendorf® 022628124 (NA)
parameter
-9-40 °C temp. range
100-4,400 rpm speed
3,000 × g max. RCF
technique(s)
centrifugation: suitable
W × H × D
38 cm × 58 cm × 26 cm
weight
36 kg
Looking for similar products? Visit Product Comparison Guide
General description
The right model for your application:
- Centrifuge 5702 for standard applications
- Refrigerated Centrifuge 5702 R for heat sensitive samples
- Heated and refrigerated Centrifuge 5702 RH allows centrifugation under biological conditions to improve cell viability for more accurate results in downstream applications (e.g. cell cultivation)
Ergonomics
- Very compact footprint to fit on every lab bench
- Very quiet operation to benefit your environment
- Low access height for simple loading and unloading of samples
Temperature management
- Optimizes air flow minimizes sample warming during run (5702)
- FastTemp function for fast precooling (5702 R/RH)
- Active heating guarantees high temperature accuracy during the entire centrifugation cycle (only 5702 RH)
- ECO shut–off engages after 8 hours of nonuse to reduce energy consumption and extend compressor life (5702 R/RH)Centrifuge 5702/ 5702 R/5702 RH
Quality and safety
- Stainless steel rotor chamber is rust-free and easy to clean
- Imbalance detection for maximum safety
- Key lock to prevent accidental parameter changes
- Aerosol–tight caps for safe centrifugation of hazardous samples optionally available
Application
- Centrifuge 5702 for standard applications
- Refrigerated Centrifuge 5702 R for heat sensitive samples
- Heated and refrigerated Centrifuge 5702 RH allows centrifugation to improve cell viability for more accurate results in downstream applications
Features and Benefits
- Max. capacity: 4 × 100 mL
- Max. speed: 3,000 × g (4,400 rpm)
- Very compact footprint to fit on every lab bench
- Very quiet operation to benefit your environment
- Low access height for simple loading and unloading of samples
- SOFT brake function for slow acceleration and braking. Optimized for cell separation by gradient centrifugation
- “At set RPM” function starts the timer when the selected rotational speed is reached for reproducible centrifugation runs
- Key lock to prevent unintentional adjustments
- Stainless steel rotor chamber is rust-free and easy to clean
- Electronic imbalance detection for maximum safety
Special features for Centrifuge 5702 R and Centrifuge 5702 RH
- Temperature settings from -9 to 40 deg. C
- FastTemp pro® function for fast precooling
- Continuous cooling maintains a constant temperature after the run has been completed – your samples stay cool
- ECO shut-off engages after 8 hours of non-use to reduce energy consumption and extend compressor life
- Two program keys to store routine procedures
- Active heating guarantees high temperature accuracy during the entire centrifugation cycle (only 5702 RH)
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service