Skip to Content
Merck
All Photos(1)

Key Documents

EP0030077512

Eppendorf® ep Dualfilter T.I.P.S.®

10 racks x 96 ea, PCR clean, capacity 0.1-10 μL, sterile

Synonym(s):

pipetting tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
capacity:
0.1-10 μL
maximum volume:
10 μL
sterility:
sterile
feature:
DNase free
RNase free
filter
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Eppendorf® 0030077512
Pricing and availability is not currently available.

material

gray (Tray)

sterility

sterile

feature

DNase free
RNase free
filter

packaging

pack of 960 ea (10 racks of 96 tips)

manufacturer/tradename

Eppendorf® 0030077512

technique(s)

PCR: suitable

capacity

0.1-10 μL

maximum volume

10 μL

Looking for similar products? Visit Product Comparison Guide

General description

  • Eppendorf® ep Dualfilter T.I.P.S.® (0.1-10 μL) are original, supreme quality Eppendorf dual-filter pipette tips with low retention property that are uncoated, free of additives and packed in resealable bags.
  • Ultrahomogenous surface for maximum reproducibility.
  • Ultrahydrophobic surface for minimal sample loss.
  • Significantly reduced foam formation during pipetting.
  • Extreme resistance to chemicals.
  • Available in PCR clean and Eppendorf Quality.
  • Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile.
  • Sterile and pyrogen-free.

Application

  • Cell culture (media).
  • Genomics: PCR, RT-PCR, qPCR and all other types of PCR.
  • Enzymatic reactions (restriction decomposition, ligation).
  • Isolation and purification of nucleic acid.
  • Gel electrophoresis (e.g., prefabricated DNA ladders) typical detergents: SDS, Triton, X-100, Brij® 35, Tween® 20, CHAPS.
  • Proteomics (all types of protein examinations).
  • Protein isolation and purification.
  • NGS library preparation.

Legal Information

Brij is a registered trademark of Croda International PLC
Eppendorf is a registered trademark of Eppendorf AG
TWEEN is a registered trademark of Croda International PLC
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service