Skip to Content
Merck
All Photos(1)

Key Documents

EHU115601

Sigma-Aldrich

MISSION® esiRNA

targeting human KRT19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGCAACGAGAAGCTAACCATGCAGAACCTCAACGACCGCCTGGCCTCCTACCTGGACAAGGTGCGCGCCCTGGAGGCGGCCAACGGCGAGCTAGAGGTGAAGATCCGCGACTGGTACCAGAAGCAGGGGCCTGGGCCCTCCCGCGACTACAGCCACTACTACACGACCATCCAGGACCTGCGGGACAAGATTCTTGGTGCCACCATTGAGAACTCCAGGATTGTCCTGCAGATCGACAATGCCCGTCTGGCTGCAGATGACTTCCGAACCAAGTTTGAGACGGAACAGGCTCTGCGCATGAGCGTGGAGGCCGACATCAACGGCCTGCGCAGGGTGCTGGATGAGCTGACCCTGGCCAGGACCGACCTGGAGATGCAGATCGAAGGCCTGAAGGAAGAGCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takayuki Kawai et al.
Cancer medicine, 6(11), 2531-2540 (2017-10-02)
The current lack of an easily measurable surrogate marker of cancer stem cells (CSCs) prevents the clinical application of CSCs for hepatocellular carcinoma (HCC). We previously reported that keratin 19 (K19) is a novel HCC-CSC marker associated with transforming growth
Masato Takano et al.
BMC cancer, 16(1), 903-903 (2016-11-20)
Keratin (K) 19-positive hepatocellular carcinoma (HCC) is well known to have a higher malignant potential than K19-negative HCC: However, the molecular mechanisms involved in K19-mediated progression of HCC remain unclear. We attempted to clarify whether K19 directly affects cell survival
Tomoaki Ohtsuka et al.
Scientific reports, 6, 39557-39557 (2016-12-23)
HER2 is a receptor tyrosine kinase and its upregulation via activating mutations or amplification has been identified in some malignant tumors, including lung cancers. Because HER2 can be a therapeutic target in HER2-driven malignancies, it is important to understand the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service