AFRC IAH-CC62
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
Mouse x Mouse Hybridoma
packaging
tube of 5 μg 99011334-DNA-5UG
pkg of vial of cells 99011334-1VL
ImmunoDon
BALB/c mouse spleen
Myeloma
NS-1
Reactivity
Anti-CD26 (formerly WC10), bovine
technique(s)
cell culture | mammalian: suitable
isotype
IgG2b
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse x Mouse hybridoma
Cell Line Description
The hybridoma IAH-CC62 secretes antibodies reactive with bovine CD26 - this was formerly known as WC10. This non-lineage specific antigen is present on bovine leukocytes and has a MW of 115 kDa and 39kDa. Cells have been grown on mouse macrophage feeder cells using hypoxanthine and thymidine at concentractions appropriate for hybridomas. It is recommended to continue this until sufficient cells have been frozen.
Immunogen
Bovine thymocytes
Application
Immunohistology, Flow cytometry, Immunoprecipation
Culture Medium
RPMI 1640 + 2 mM glutamine + HT supplement + 10% FBS
Subculture Routine
Maintain cultures between 3-9x 100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service