Skip to Content
Merck
All Photos(1)

Documents

RBL-1

86061001, rat blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

RBL-1, 86061001

biological source

rat blood

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Rat basophilic leukaemia granulocyte

Cell Line Description

Exhibits marked variations in differentiation when monitored for basophilic granule development, histamine content and membrane receptor sites for IgE.

Application

Differentiation studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1mM Sodium Pyruvate (NaP) + 1% Non Essential Amino Acids (NEAA) + 10-20% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3x100,000 to 1x1,000,000 cells/ml; 5% CO2; 37°C. Treat as suspension when sub-culturing. Some fibroblast-like cells within the population will attach. Please disregard these and subculture as per normal suspension cell lines.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service