Skip to Content
Merck
All Photos(1)

Key Documents

HEK293A GFP-LC3

14050801, human kidney, Small slightly rounded cells growing in a dispersed manner

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

HEK293A GFP-LC3, 14050801

biological source

human kidney

description


Keywords: Human embryonic kidney, autophagy, HEK293, EGFP-rat LC3, reporter cell line, ATG8, microtubule-associated protein 1 light chain 3 beta (MAP1LC3B)

growth mode

Adherent

morphology

Small slightly rounded cells growing in a dispersed manner

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Description

A stable cell line expressing EGFP-tagged LC3 (rat) in background of HEK293 cell line. This reporter cell line can be used to monitor induction of autophagy after amino acid starvation or rapamycin treatment as well as autophagosome formation. Also, this cell line provides a useful tool for the study of autophagy using GFP-LC3 as a standard assay read out by both immunofluorescence and biochemical methods.

To generate the cell line EGFP-LC3 (rat) plasmid was transfected into HEK293A cells. The cells were selected with 800ug/ml Geneticin. Single cell clones were selected and grown up. Several clones were tested and clone 2GL9 was taken as the best based on a number of parameters (including GFP-LC3 expression levels and responsiveness to amino acid deprivation).

Description of the genetic modification to generate this cell line: HEK293 cells transfected with a GFP-LC3 (green fluorescent protein-microtubule-associated protein 1 light chain 3) plasmid. cDNA encoding rat LC3 was obtained by RT–PCR from rat brain total cDNA and subcloned into the EcoRI site of the eukaryotic expression vector pCI-neo (Promega, Madison, WI). It was then excised and the LC3 cDNA inserted into the BglII and EcoRI sites of pEGFP-C1, a GFP fusion protein expression vector (Clontech Laboratories), Kabeya Y, et.al. (2000).

Culture Medium

Dulbecco′s Modified Eagle′s Medium, high glucose (DMEM) + 2mM Glutamine + 10% Fetal Bovine Serum (FBS).

Geneticin at (400ug/ml) can be added for maintenance but this is not normally necessary as the GFP-LC3 expression level is maintained over at least 20 passages.

Subculture Routine

Trypsinise cultures at 80% confluence with 0.05% Trypsin/EDTA. Seed flasks at 2 x 104 cells/ cm2. Cultures must be incubated in a humidified 10% CO2/90% air incubator at 37oC.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service