Skip to Content
Merck
All Photos(1)

Documents

C11100P

Sigma-Aldrich

DMT-dC(bz) Pharmadite®

Synonym(s):

DMT-dC(bz) Phosphoramidite, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxycytidine, 3′-[2-cyanoethyl N,N-?bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H52N5O8P
Molecular Weight:
833.91
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.51

biological source

non-animal source (no BSE/TSE risk)

Quality Level

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥99.5% (reversed phase HPLC)
>99.5% (31P-NMR)

form

powder or granules

technique(s)

oligo synthesis: suitable

impurities

>= 170ppm, none detected (S/N <2.5)
≤0.1% P(III) 100-169ppm (31P-NMR)
≤0.1% single unspecified Imprity (reversed phase HPLC)
≤0.3% dC2 (reversed phase HPLC, DMT-dC(bz) cyanoethyl-H-posphonate)
≤0.3% dC3 (reversed phase HPLC, DMT-dC(bz) phosphoramidate)
≤0.3% dC5 (reversed phase HPLC, O3′-Benzoyl-O5′-DMT-dC(bz))
≤0.30 wt. % water content (Karl Fischer)
≤0.5% P(V) -25-99ppm (31P-NMR)
≤3.0 wt. % residual solvent content

color

white to off-white

solubility

soluble, clear

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

nucleoside profile

base: deoxycytidine
base protecting group: benzoyl
2' protecting group: none
5' protecting group: DMT
deprotection: standard

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)N3C=CC(NC(=O)c4ccccc4)=NC3=O)(c5ccccc5)c6ccc(OC)cc6

InChI

1S/C46H52N5O8P/c1-32(2)51(33(3)4)60(57-29-13-27-47)59-40-30-43(50-28-26-42(49-45(50)53)48-44(52)34-14-9-7-10-15-34)58-41(40)31-56-46(35-16-11-8-12-17-35,36-18-22-38(54-5)23-19-36)37-20-24-39(55-6)25-21-37/h7-12,14-26,28,32-33,40-41,43H,13,29-31H2,1-6H3,(H,48,49,52,53)/t40-,41+,43+,60?/m0/s1

InChI key

PGTNFMKLGRFZDX-SALLYJDFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DMT-dC(bz) Pharmadite® is a Pharmadite® product line that represents a new class of standard protected DNA phosphoramidites designed with highly controlled impurity profiles and exceptional overall purity.Pharmadite phosphoramidites are characterized by their exceptional purity and well-defined impurity profile. In particular, they are essentially free fromcontaminants that interfere in coupling reactions, such as other nucleosidic or non-nucleosidic phosphoramidites (P(III)-contaminants). All remainingimpurities in Pharmadite phosphopramidites, if present, are identified, characterized, and quantified. The impact of any such defined impurities on the synthesis of oligonucleotides is well understood and has been shown to be insignificant.

Legal Information

Pharmadite is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service