Key Documents
BR31571
BRAND® BLAUBRAND® measuring cylinder with serialized numbers
capacity 10 mL
About This Item
UNSPSC Code:
41121800
packaging
pkg of 12 ea
manufacturer/tradename
BRAND 27452
H
115 mm
capacity
10 mL
General description
Measuring cylinders tall form. Class A DE-M marking to DIN EN ISO 4788. Made from DURAN(R), calibrated to contain (TC, In). With spout and hexagonal glass base. Ring marks at major graduations. High contrast blue enamel graduations and inscription
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)