Key Documents
BAF134730001
Bel-Art® Write-on Tape Dispenser
Wall Mount Adapter for Economy Model
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F13473-0001
L × W
100 mm × 125 mm
General description
Use to mount the dispenser vertically or horizontally.
U-shaped stainless steel foot positions ′Write-On Label Tape Dispenser at a 45° angle off of the wall for improved ergonomics.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Write-on is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)