Skip to Content
Merck
All Photos(1)

Documents

AXYT300

Pipette tips, for P2/P10

size 0.5-10 μL, clear, pkg of 20x1000tips/cs

Synonym(s):

Pipette tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02

material

clear

sterility

non-sterile

feature

DNA free
RNase free
barrier: no
endotoxin free
filter: no

manufacturer/tradename

Axygen Inc T-300
Corning

packaging

pkg of 20x1000tips/cs

size

0.5-10 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

  • Axygen® microvolume pipette tips are made of polypropylene.
  • The tips are optimized for use with both single and multi-channel pipettors.
  • Features a flexible wall and internal sealing ring for a secure fit.

Other Notes

Browse all pipette tips

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service