Skip to Content
Merck
All Photos(1)

Documents

A0355

Sigma-Aldrich

AC-7954

≥98% (HPLC), solid

Synonym(s):

(±)-3-(4-Chlorophenyl)-3-(2-(dimethylamino)ethyl)isochroman-1-one hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20ClNO2 · HCl
CAS Number:
Molecular Weight:
366.28
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

white

solubility

H2O: ~28 mg/mL

SMILES string

Cl.CN(C)CCC1(Cc2ccccc2C(=O)O1)c3ccc(Cl)cc3

InChI

1S/C19H20ClNO2.ClH/c1-21(2)12-11-19(15-7-9-16(20)10-8-15)13-14-5-3-4-6-17(14)18(22)23-19;/h3-10H,11-13H2,1-2H3;1H

InChI key

AJPGLOWFIBOGMH-UHFFFAOYSA-N

Gene Information

human ... UTS2R(2837)

Biochem/physiol Actions

Nonpeptide GPR14 / urotensin-UII (UII) receptor agonist that has an EC50 of 300 nM at the human UII receptor (h-UTR). AC-7954 has been shown to exist in two conformations, A and B; both conformations have a pharmacophoric distance within 0.5 Å of P5U, a potent peptidic UII agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glenn E Croston et al.
Journal of medicinal chemistry, 45(23), 4950-4953 (2002-11-01)
A functional cell-based screen identified 3-(4-chlorophenyl)-3-(2-(dimethylamino)ethyl)isochroman-1-one hydrochloride (AC-7954, 1) as a nonpeptidic agonist of the urotensin-II receptor. Racemic 1 had an EC50 of 300 nM at the human UII receptor and was highly selective. Testing of the enantiopure (+)- and
Alfonso Carotenuto et al.
Journal of medicinal chemistry, 47(7), 1652-1661 (2004-03-19)
Urotensin II (U-II) is a disulfide-bridged undecapeptide recently identified as the ligand of an orphan G-protein-coupled receptor. Human U-II (H-Glu-Thr-Pro-Asp-cyclo[Cys-Phe-Trp-Lys-Tyr-Cys]-Val-OH) has been described as the most potent vasoconstrictor compound identified to date. With the aim of elucidating the active conformation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service