21818
Carbol-Fuchsin Dye Powder
for microscopy
Synonym(s):
Parafuchsin + phenol
About This Item
Recommended Products
grade
for microscopy
form
solid
InChI
1S/C21H21N3.ClH/c1-13-11-16(5-9-19(13)23)21(15-3-7-18(22)8-4-15)17-6-10-20(24)14(2)12-17;/h3-12,22H,23-24H2,1-2H3;1H
InChI key
YCIVZWKFFBDLTG-UHFFFAOYSA-N
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2
Target Organs
Nervous system,Kidney,Liver,Skin
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service