Skip to Content
Merck
All Photos(1)

Key Documents

VT000249

Supelco

Pseudomonas aeruginosa WDCM 00024 Vitroids

variable CFU mean value range, certified reference material, suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

−20°C

suitability

Pseudomonas spp.

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Download your certificate at: http://www.sigma-aldrich.com.

Please specify if you require a single batch - in the remark field when ordering (only for orders with more than 15 packs)

Other Notes

ATCC® 10145, NCTC 10332, NCPF -, DSM 50071, LMG 1242, IHEM -, CIP 76.110
Origin Strain No. CECT® 110

Legal Information

Derived from CECT® strains
ATCC is a registered trademark of American Type Culture Collection
CECT is a registered trademark of Universitat de Valencia
Vitroids is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service