Skip to Content
Merck
All Photos(1)

Key Documents

45532

Supelco

Indole-3-butyric acid

PESTANAL®, analytical standard

Synonym(s):

4-(3-Indolyl)butanoic acid, 4-(3-Indolyl)butyric acid, IBA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13NO2
CAS Number:
Molecular Weight:
203.24
Beilstein:
171120
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)CCCc1c[nH]c2ccccc12

InChI

1S/C12H13NO2/c14-12(15)7-3-4-9-8-13-11-6-2-1-5-10(9)11/h1-2,5-6,8,13H,3-4,7H2,(H,14,15)

InChI key

JTEDVYBZBROSJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indole-3-butyric acid is a plant hormone in the auxin family and is widely used in agriculture since it induces rooting in various plants species such as mung bean (Vigna radiata L.) cuttings.

Application

Indole-3-butyric acid may be used as a reference standard in the determination of indole-3-butyric acid in mung bean sprouts using high performance liquid chromatography with chemiluminescence (CL) detection.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic analysis of indole-3-butyric acid responses in Arabidopsis thaliana reveals four mutant classes.
Zolman KB, et al.
Genetics, 156(3), 1323-1337 (2000)
Comparison of movement and metabolism of indole-3-acetic acid and indole-3-butyric acid in mung bean cuttings.
Wiesman Z, et al.
Physiologia Plantarum, 74(3), 556-560 (1988)
Determination of indole-3-acetic acid and indole-3-butyric acid in mung bean sprouts using high performance liquid chromatography with immobilized ru (bpy) 32+-kmno4 chemiluminescence detection.
Xi Z, et al.
Talanta, 79(2), 216-221 (2009)
Lucia C Strader et al.
The Plant cell, 23(3), 984-999 (2011-03-17)
Levels of auxin, which regulates both cell division and cell elongation in plant development, are controlled by synthesis, inactivation, transport, and the use of storage forms. However, the specific contributions of various inputs to the active auxin pool are not
Kang Wei et al.
Gene, 514(2), 91-98 (2012-12-04)
The plant hormone auxin plays a key role in adventitious rooting. To increase our understanding of genes involved in adventitious root formation, we identified transcripts differentially expressed in single nodal cuttings of Camellia sinensis treated with or without indole-3-butyric acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service