Skip to Content
Merck
All Photos(1)

Key Documents

31553

Supelco

Chlorpyrifos solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11Cl3NO3PS
CAS Number:
Molecular Weight:
350.59
Beilstein:
1545756
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)Oc1nc(Cl)c(Cl)cc1Cl

InChI

1S/C9H11Cl3NO3PS/c1-3-14-17(18,15-4-2)16-9-7(11)5-6(10)8(12)13-9/h5H,3-4H2,1-2H3

InChI key

SBPBAQFWLVIOKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qinghai Wang et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 67(9), 1908-1915 (2013-05-10)
For understanding the influence of initial concentrations of pesticides in the water body on removal efficiency of the contaminant by aquatic plants, one hydroponics experiment was used to investigate the influence of initial concentration (1-16 mg L(-1)) on toxicity and
Mansur A Sandhu et al.
The Journal of toxicological sciences, 38(2), 237-244 (2013-03-29)
The oral intubation of chlorpyrifos, an extensively used organophosphate insecticide, was tested for its capability to induce in vivo genotoxic upshot in blood lymphocytes of 24 male and female Wistar rats using biomarker of genotoxicity. Rats were orally administered with
Hyeong-Moo Shin et al.
Environmental science & technology, 47(2), 859-867 (2012-12-19)
Indoor residence times of semivolatile organic compounds (SVOCs) are a major and mostly unavailable input for residential exposure assessment. We calculated residence times for a suite of SVOCs using a fugacity model applied to residential environments. Residence times depend on
W Guo et al.
Journal of economic entomology, 106(3), 1112-1117 (2013-07-20)
The efficacy of entomopathogenic nematodes (EPNs) against the white grub Holotrichia parallela Motschulsky (Coleoptera: Scarabaeidae) in the peanut (Arachis hypogaea L.) fields was evaluated. In the first experiment with a high initial white grub population (24.65 +/- 2.44 larvae per
Robert U Svensson et al.
Journal of environmental pathology, toxicology and oncology : official organ of the International Society for Environmental Toxicology and Cancer, 32(1), 29-39 (2013-06-14)
Environmental factors are likely to interact with genetic determinants to influence prostate cancer progression. The Agricultural Health Study has identified an association between exposure to organophosphorous pesticides including chlorpyrifos, and increased prostate cancer risk in pesticide applicators with a first-degree

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service